• USA Home
  • 34420 - α,α′-Dibromo-o-xylene

EMAIL THIS PAGE TO A FRIEND
34420 Sigma-Aldrich

α,α′-Dibromo-o-xylene

purum, ≥97.0% (AT)

Synonym: 1,2-Bis(bromomethyl)benzene, o-Xylylene dibromide

  • CAS Number 91-13-4

  • Linear Formula C6H4(CH2Br)2

  • Molecular Weight 263.96

  •  Beilstein Registry Number 637159

  •  EC Number 202-042-7

  •  MDL number MFCD00000175

  •  PubChem Substance ID 329754740

Purchase

Properties

Related Categories Aryl, Building Blocks, C8, Chemical Synthesis, Halogenated Hydrocarbons,
InChI Key   KGKAYWMGPDWLQZ-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (AT)
mp   91-94 °C(lit.)
  91-94 °C
density   1.96 g/mL at 25 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3448 6.1 / PGII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

D44405 α,α′-Dibromo-o-xylene, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?