• USA Home
  • 34430 - α,α′-Dibromo-m-xylene

EMAIL THIS PAGE TO A FRIEND
34430 Sigma-Aldrich

α,α′-Dibromo-m-xylene

purum, ≥95.0% (GC)

Synonym: 1,3-Bis(bromomethyl)benzene, m-Xylylene dibromide

  • CAS Number 626-15-3

  • Linear Formula C6H4(CH2Br)2

  • Molecular Weight 263.96

  •  Beilstein Registry Number 971085

  •  EC Number 210-931-6

  •  MDL number MFCD00000178

  •  PubChem Substance ID 24861241

Purchase

Properties

Related Categories Aryl, Building Blocks, C8, Chemical Synthesis, Halogenated Hydrocarbons,
InChI Key   OXHOPZLBSSTTBU-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
bp   135-140 °C/20 mmHg(lit.)
mp   74-77 °C(lit.)
  74-77 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3448 6.1 / PGII
WGK Germany 
3
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

125911 α,α′-Dibromo-m-xylene, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?