346144 Sigma-Aldrich

Diethyl ether

reagent grade, ≥98.0%, contains ~2% ethanol and ~10ppm BHT as inhibitor

DOWNLOAD MSDS (PDF)

Synonym: Ether, Ethyl ether

  • CAS Number 60-29-7

  • Linear Formula (CH3CH2)2O

  • Molecular Weight 74.12

  •  Beilstein Registry Number 1696894

  •  EC Number 200-467-2

  •  MDL number MFCD00011646

  •  PubChem Substance ID 24861375

Properties

Related Categories ACS and Reagent Grade Solvents, Diethyl Ether, Reagent Grade Solvents, Solvent by Type, Solvent Bottles,
grade   reagent grade
vapor density   2.6 (vs air)
vapor pressure   28.5 psi ( 55 °C)
  8.38 psi ( 20 °C)
assay   ≥98.0%
autoignition temp.   320 °F
  356 °F
contains   ~2% ethanol and ~10ppm BHT as inhibitor
expl. lim.   36.5 %
refractive index   n20/D 1.3530(lit.)
bp   34.6 °C(lit.)
mp   −116 °C(lit.)
density   0.706 g/mL at 25 °C(lit.)

Description

Packaging

1 L in Sure/Seal™

Price and Availability

Customers Also Viewed

Diethyl ether

contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%

Diethyl ether

ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor

Diethyl ether

ACS reagent, ≥98.0%, contains ~2% ethanol and ~10ppm BHT as inhibitor

Diethyl ether

reagent grade, contains 1 ppm BHT as stabilizer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?