• USA Home
  • 34615 - Polynuclear aromatic hydrocarbons 6 solution

EMAIL THIS PAGE TO A FRIEND
34615 Sigma-Aldrich

Polynuclear aromatic hydrocarbons 6 solution

analytical standard, in cyclohexane

Purchase

Properties

grade   analytical standard
description   suitable for TrinkwV (BRD) Method
quality   in cyclohexane
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   multi-component solution
storage temp.   2-8°C

Description

Components

1 μl contains
10 ng fluoranthene and 2 ng each benzo[b]fluoranthene, benzo[k]fluoranthene, benzo[a]pyrene, benzo[ghi]perylene and indeno[1,2,3-cd]pyrene

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1145 3/PG 2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?