• USA Home
  • 34632 - Mercury(II) sulfate-Sulfuric acid solution

EMAIL THIS PAGE TO A FRIEND
34632 Sigma-Aldrich

Mercury(II) sulfate-Sulfuric acid solution

for COD determination according to DIN 38409, part 41, 5.855-5.913 g/L K2Cr2O7 basis

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Waste Water
grade   for COD determination according to DIN 38409, part 41
InChI Key   DOBUSJIVSSJEDA-UHFFFAOYSA-L
assay   5.855-5.913 g/L K2Cr2O7 basis
concentration   80 g/L in sulfuric acid

Description

Caution

Restricted to professional users. Attention − Avoid exposure − obtain special instructions before use.

Other Notes

Same product — same quality — now Sigma-Aldrich brand

General description

Addition of acetylene, to mercury (II) sulphate in sulphuric acid solution, results in production of a series of products; acetylide (HgC2), organometallic compounds (C2H2HgCl2.HgCl), chlorovinylmercury, aldehydes and ketones and mercuration products of aldehydes and ketones.

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN 2922 6.1(8) / PGII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?