• USA Home
  • 34687 - Di-tert-butyl N,N-diisopropylphosphoramidite

EMAIL THIS PAGE TO A FRIEND
34687 Sigma-Aldrich

Di-tert-butyl N,N-diisopropylphosphoramidite

technical, ~90% (GC/31P-NMR)

  • CAS Number 137348-86-8

  • Linear Formula [(CH3)2CH]2NP[OC(CH3)3]2

  • Molecular Weight 277.38

  •  Beilstein Registry Number 4858847

  •  MDL number MFCD00153506

  •  PubChem Substance ID 329755053

Purchase

Properties

InChI Key   YGFLCNPXEPDANQ-UHFFFAOYSA-N
grade   technical
assay   ~90% (GC/31P-NMR)
refractive index   n20/D 1.444(lit.)
bp   85-90 °C/0.2 mmHg(lit.)
density   0.879 g/mL at 25 °C(lit.)
shipped in   wet ice
storage temp.   −20°C

Description

Other Notes

Phosphitylating agent for synthesizing derivatives of O4-phospho-tyrosine

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

419362 Di-tert-butyl N,N-diisopropylphosphoramidite, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?