EMAIL THIS PAGE TO A FRIEND
34717 Sigma-Aldrich

AQUANAL GH

for determination of total water hardness

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Kits/Cases, Potable- & Waste-Water Testing
grade   for determination of total water hardness
impurities   N,N-dimethyl-1,3-diaminopropane

Description

General description

Aquanal kits can be used for the determination of hardness and phosphate ammonia, nitrate, and nitrite concentrations.

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?