EMAIL THIS PAGE TO A FRIEND
34718 Sigma-Aldrich

AQUANAL SV

for determination of acid consumption in water

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Kits/Cases, Potable- & Waste-Water Testing
grade   for determination of acid consumption in water

Description

General description

Aquanal kits can be used for the determination of hardness and phosphate ammonia, nitrate, and nitrite concentrations.

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN3289 - class 6.1 - PG 2 - acidic - Toxic liquid, corrosive, inorganic, n.o.s., HI: all
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?