• USA Home
  • 34779 - N,N-Dibutyl-L-norephedrine

EMAIL THIS PAGE TO A FRIEND
34779 Sigma-Aldrich

N,N-Dibutyl-L-norephedrine

purum, ≥95.0% (sum of enantiomers, GC)

Synonym: (+)-DBNE, (1R,2S)-2-Dibutylamino-1-phenyl-1-propanol

  • CAS Number 115651-77-9

  • Empirical Formula (Hill Notation) C17H29NO

  • Molecular Weight 263.42

  •  Beilstein Registry Number 5337383

  •  MDL number MFCD00142689

  •  PubChem Substance ID 329755056

Purchase

Properties

InChI Key   BRRGNOFUBFINSX-RDJZCZTQSA-N
grade   purum
assay   ≥95.0% (sum of enantiomers, GC)
optical activity   [α]20/D +24±1°, c = 2% in hexane
refractive index   n20/D 1.410
density   0.944 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

407135 (1R,2S)-(+)-2-(Dibutylamino)-1-phenyl-1-propanol, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?