34851 Sigma-Aldrich

Acetonitrile

CHROMASOLV® gradient grade, for HPLC, ≥99.9%

DOWNLOAD MSDS (PDF)

Synonym: ACN, Methyl cyanide

Properties

Related Categories 3-MCPD, Acetonitrile and Acetonitrile Solutions, Acrylamide, Adulterants, Allergens,
grade   CHROMASOLV® gradient grade
  for HPLC
vapor density   1.41 (vs air)
vapor pressure   72.8 mmHg ( 20 °C)
assay   ≥99.9%
autoignition temp.   973 °F
expl. lim.   16 %
measuring range   ≤15 mAU baseline drift, (210 nm)
impurities   ≤0.0002% free alkali (as NH3)
  ≤0.0005% non-volatile matter
  ≤0.001% free acid (as CH3COOH)
  ≤0.02% water (Karl Fischer)
  ≤0.5 ppb fluorescence (quinine) at 365 nm
  ≤1 ppb fluorescence (quinine) at 254 nm
refractive index   n20/D 1.344(lit.)
bp   81-82 °C(lit.)
mp   −48 °C(lit.)
density   0.786 g/mL at 25 °C(lit.)
abs.   HPLC-gradient/210 nm ≤3 mAU
  HPLC-gradient/254 nm ≤0.5 mAU
UV absorption   λ: 195 nm Amax: ≤0.12
  λ: 200 nm Amax: ≤0.032
  λ: 230 nm Amax: ≤0.0044
  λ: 235 nm Amax: ≤0.0044
  λ: 250 nm Amax: ≤0.0044
  λ: 400 nm Amax: ≤0.0044
storage temp.   room temp

Description

Frequently Asked Questions

Live Chat and Frequently Asked Questions are available for this Product.

Other Notes

Pure-Pac® II containers require the Micromatic MacroValve coupler for dispensing solvents, Z560723.

Packaging

1 pkg in Pure-Pac™ 2

100 mL in glass bottle

200 L in Pure-Pac™ 2

1, 6×1, 144×1, 2, 4×2, 4, 4×4 L in glass bottle

18 L in Pure-Pac™ 1

20 L in Nowpak™

20, 50 L in Pure-Pac™ 2

56, 200 L in Pure-Pac™ 1

7 L in ss drum

View returnable container options.

Preparation Note

Product filtered through a 0.2 μm filter

Legal Information

CHROMASOLV is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH

Pure-Pac is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Customers Also Viewed

Acetonitrile

LC-MS CHROMASOLV®

Acetonitrile

CHROMASOLV® Plus, for HPLC, ≥99.9%

Acetonitrile

CHROMASOLV® Plus, for HPLC, ≥99.9%

Acetonitrile

CHROMASOLV® gradient grade, for HPLC, ≥99.9%

Methanol

CHROMASOLV®, for HPLC, ≥99.9%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?