EMAIL THIS PAGE TO A FRIEND

34865 Sigma-Aldrich

Tetrahydrofuran

inhibitor-free, CHROMASOLV® Plus, for HPLC, ≥99.9%

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, CHROMASOLV Plus, Chromatography Reagents & Solvents, HPLC & UHPLC Solvents (CHROMASOLV),
grade   CHROMASOLV® Plus
  for HPLC
vapor density   2.5 (vs air)
vapor pressure   114 mmHg ( 15 °C)
  143 mmHg ( 20 °C)
assay   ≥99.9%
autoignition temp.   610 °F
expl. lim.   11.8 %
impurities   ≤0.0005% non-volatile matter
  ≤0.002% free acid (as CH3COOH)
  ≤0.05% peroxides (as H2O2)
  ≤0.05% water (Karl Fischer)
evapn. residue   ≤0.0005%
color   APHA: ≤10
refractive index   n20/D 1.407(lit.)
bp   65-67 °C(lit.)
mp   −108 °C(lit.)
density   0.889 g/mL at 25 °C(lit.)
absorption   absorption/212 nm ≤1.0
  absorption/245 nm ≤0.26
  absorption/250 nm ≤0.18
  absorption/275 nm ≤0.046
  absorption/300 nm ≤0.02
  absorption/315 nm ≤0.0044
  absorption/350 nm ≤0.005
  absorption/400 nm ≤0.005
UV absorption   λ: 245 nm Amax: ≤0.26
  λ: 275 nm Amax: ≤0.046
  λ: 315 μm Amax: ≤0.0044
suitability   complies for IR spectroscopy

Description

Application

Suitable for HPLC, spectrophotometry, environmental testing

Packaging

1, 6×1, 2, 4, 4×4 L in glass bottle

20 L in Nowpak™

100, 12×100 mL in glass bottle

200 L in Pure-Pac™ 1

Legal Information

CHROMASOLV is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH

Price and Availability

Timestrip - Time & Temperature Indicators


Timestrip
Discover how Timestrip® indicators will help you monitor expiry dates or excessive temperatures.




Greener/Safer Solvent Dispensing Options


Returnable ContainersOur wide selection of solvents are
available in returnable containers with
safe and efficient dispensing options that meet your application needs.




Additional Products to Explore

Dichloromethane

anhydrous, ≥99.8%, contains 50-150 ppm amylene as stabilizer

Diethyl ether

contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%

Methanol
322415Methanol

anhydrous, 99.8%

<I>N,N</I>-Dimethylformamide

anhydrous, 99.8%

Toluene
244511Toluene

anhydrous, 99.8%

Customers Also Viewed

Tetrahydrofuran

anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%

Tetrahydrofuran

CHROMASOLV® Plus, for HPLC, ≥99.9%, inhibitor-free

Tetrahydrofuran

contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
May form explosive peroxides.
Hazard Codes (Europe) 
Risk Statements (Europe) 
Safety Statements (Europe) 
13-16-29-33-36-37-46
RIDADR 
UN 2056 3 / PGII
WGK Germany 
1
RTECS 
LU5950000
Flash Point(F) 
1.4 °F
Flash Point(C) 
-17 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Determination of Water Content in Tetrahydrofuran Using Karl Fischer Titration

Ethers behave similarly to hydrocarbons. They are mostly adequately soluble in methanol. Only long chain ethers require the addition of propanol or chloroform as solubilizers. Double bonds can someti...
Keywords: Solvents, Titrations

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Kinetics of 1,4-dioxane biodegradation by monooxygenase-expressing bacteria. Mahendra S and Alvarez-Cohen L Environ. Sci. Technol. 40(17), 5435-42, (2006)

Loading...


Enhanced mobility of fullerene (C60) nanoparticles in the presence of stabilizing agents. Wang Y, Li Y, Costanza J, et al. Environ. Sci. Technol. 46(21), 11761-9, (2012)

Loading...


Total, elastic, and inelastic cross sections for positron and electron collisions with tetrahydrofuran. Chiari L, Anderson E, Tattersall W, et al. J. Chem. Phys. 138(7), 074301, (2013)

Loading...


Reduced bacterial adhesion to fibrinogen-coated substrates via nitric oxide release. Charville GW Biomaterials 29(30), 4039-44, (2008)

Loading...


Hollow interior structure of spin-coated polymer thin films revealed by ToF-SIMS three-dimensional imaging. Ren X, Weng LT, Chan CM, et al. Anal. Chem. 84(20), 8497-504, (2012)

Loading...


Designing secondary structures: 5-azidomethyl tetrahydrofuran-2-carboxylates as carbohydrate-derived dipeptide isosteres. Smith MD and Fleet GW J. Pept. Sci. 5(10), 425-41, (1999)

Loading...


Site-isolated redox reactivity in a trinuclear iron complex. Eames EV and Betley TA Inorg. Chem. 51(19), 10274-8, (2012)

Loading...


Weak coordination of neutral S- and O-donor proximal ligands to a ferrous porphyrin nitrosyl. Characterization of 6-coordinate complexes at low T. Martirosyan GG, Kurtikyan TS, Azizyan AS, et al. J. Inorg. Biochem. 121, 129-33, (2013)

Loading...


The development of a method to quantify encapsulated and free prednisolone phosphate in liposomal formulations. Smits EA, Smits CJ, and Vromans H J. Pharm. Biomed. Anal. 75, 47-54, (2013)

Loading...


Glucuronidation of dihydroartemisinin in vivo and by human liver microsomes and expressed UDP-glucuronosyltransferases. Ilett KF, Ethell BT, Maggs JL, et al. Drug Metab. Dispos. 30(9), 1005-12, (2002)

Loading...


Tetrahydrofuran-containing macrolides: a fascinating gift from the deep sea. Lorente A, Lamariano-Merketegi J, Albericio F, et al. Chem. Rev. 113(7), 4567-610, (2013)

Loading...


Controlling morphology and chain aggregation in semiconducting conjugated polymers: the role of solvent on optical gain in MEH-PPV. Lampert ZE, Reynolds CL, Papanikolas JM, et al. J. Phys. Chem. B 116(42), 12835-41, (2012)

Loading...


Three-dimensional imaging of solvent-cleared organs using 3DISCO. Ertürk A, Becker K, Jährling N, et al. Nat. Protoc. 7(11), 1983-95, (2012)

Loading...


Tetrahydrofuran poisoning after occupational exposure. Garnier R, Rosenberg N, Puissant JM, et al. Br. J. Ind. Med. 46(9), 677-8, (1989)

Loading...


Comparison of the reactivity of the three distonic isomers of the pyridine radical cation toward tetrahydrofuran in solution and in the gas phase. Widjaja F, Jin Z, Nash JJ, et al. J. Am. Soc. Mass Spectrom. 24(4), 469-80, (2013)

Loading...


Chiral separations by HPLC on immobilized polysaccharide chiral stationary phases. Ali I, Al-Othman ZA, and Aboul-Enein HY Methods Mol. Biol. 970, 127-35, (2013)

Loading...


International Conference on Harmonisation; final recommendations on the revision of the permitted daily exposures for two solvents, n-methylpyrrolidone and tetrahydrofuran, according to the maintenance procedures for the guidance Q3C Impurities: Residual Solvents; Availability. Notice. Fed. Regist. 68(219), 64352-3, (2003)

Loading...


Low-energy electron scattering by cellulose and hemicellulose components. de Oliveira EM, da Costa RF, Sanchez Sd, et al. Phys. Chem. Chem. Phys. 15(5), 1682-9, (2013)

Loading...


Production of all trans-beta-carotene by using impinging flow of supercritical carbon dioxide anti-solvent pulverization. Shen YC, Chng LM, Wang YC, et al. J. Chromatogr. A 1270, 1-8, (2012)

Loading...


Direct functionalization of tetrahydrofuran and 1,4-dioxane: nickel-catalyzed oxidative C(sp3)-H arylation. Liu D, Liu C, Li H, et al. Angew. Chem. Int. Ed. Engl. 52(16), 4453-6, (2013)

Loading...


Taming of fluoroform: direct nucleophilic trifluoromethylation of Si, B, S, and C centers. Prakash GK, Jog PV, Batamack PT, et al. Science 338(6112), 1324-7, (2012)

Loading...


Synthesis and biological evaluation as microtubule-active agents of several tetrahydrofuran and spiroacetal derivatives. Carda M, Murga J, Paños J, et al. Curr. Med. Chem. 20(9), 1173-82, (2013)

Loading...


Tetrahydrofuran, tetrahydropyran, triazoles and related heterocyclic derivatives as HIV protease inhibitors. Ghosh AK and Anderson DD Future Med. Chem. 3(9), 1181-97, (2011)

Loading...


Biological monitoring of occupational exposure to tetrahydrofuran. Ong CN, Chia SE, Phoon WH, et al. Br. J. Ind. Med. 48(9), 616-21, (1991)

Loading...


Extreme multidrug resistant HIV-1 protease with 20 mutations is resistant to novel protease inhibitors with P1'-pyrrolidinone or P2-tris-tetrahydrofuran. Agniswamy J, Shen CH, Wang YF, et al. J. Med. Chem. 56(10), 4017-27, (2013)

Loading...


Diffusive hydrogen inter-cage migration in hydrogen and hydrogen-tetrahydrofuran clathrate hydrates. Cao H, English NJ, and MacElroy JM J. Chem. Phys. 138(9), 094507, (2013)

Loading...


Familial giant cell hepatitis associated with synthesis of 3 beta, 7 alpha-dihydroxy-and 3 beta,7 alpha, 12 alpha-trihydroxy-5-cholenoic acids. Clayton PT, Leonard JV, Lawson AM, et al. J. Clin. Invest. 79(4), 1031-8, (1987)

Loading...


Biodegradation of tetrahydrofuran and 1,4-dioxane by soluble diiron monooxygenase in Pseudonocardia sp. strain ENV478. Masuda H, McClay K, Steffan RJ, et al. J. Mol. Microbiol. Biotechnol. 22(5), 312-6, (2012)

Loading...


Vibrational Raman spectra of hydrogen clathrate hydrates from density functional theory. Ramya KR and Venkatnathan A J. Chem. Phys. 138(12), 124305, (2013)

Loading...


Structural basis for the recognition and cleavage of abasic DNA in Neisseria meningitidis. Lu D, Silhan J, MacDonald JT, et al. Proc. Natl. Acad. Sci. U. S. A. 109(42), 16852-7, (2012)

Loading...


Toward prediction of alkane/water partition coefficients. Anita Toulmin et al J. Med. Chem. 51, 3720-30, (2008)

Loading...


Convenient QSAR model for predicting the complexation of structurally diverse compounds with beta-cyclodextrins. Alfonso Pérez-Garrido et al Bioorg. Med. Chem. 17, 896-904, (2009)

Loading...


Metabolic disposition and pharmacokinetics of [14C]-amprenavir, a human immunodeficiency virus type 1 (HIV-1) protease inhibitor, administered as a single oral dose to healthy male subjects. Sadler BM, Chittick GE, Polk RE, et al. J. Clin. Pharmacol. 41(4), 386-96, (2001)

Loading...


Chiral recognition of amino acids by use of a fluorescent resorcinarene. GERALD I. RICHARD et al Appl. Spectroscopy 62, 476-80, (2009)

Loading...


Identification and quantitation of xenobiotics by 1H NMR spectroscopy in poisoning cases. Imbenotte M, Azaroual N, Cartigny B, et al. Forensic Sci. Int. 133(1-2), 132-5, (2003)

Loading...


Solvent effect in reactions using Stryker's reagent. Sass DC, Heleno VC, Cavalcante S, et al. J. Org. Chem. 77(20), 9374-8, (2012)

Loading...


Bis-tetrahydrofuran: a privileged ligand for darunavir and a new generation of hiv protease inhibitors that combat drug resistance. Ghosh AK, Ramu Sridhar P, Kumaragurubaran N, et al. ChemMedChem 1(9), 939-50, (2006)

Loading...


Development of QSAR models for predicting hepatocarcinogenic toxicity of chemicals. Ilaria Massarelli et al Eur. J. Med. Chem. 44, 3658-64, (2009)

Loading...


The effect of tetrahydrofuran on biological systems: does a hepatotoxic potential exist? Moody DE Drug Chem. Toxicol. 14(4), 319-42, (1991)

Loading...


Simultaneous determination of vitamins C, E and beta-carotene in human plasma by high-performance liquid chromatography with photodiode-array detection. Zhao B, Tham SY, Lu J, et al. J. Pharm. Pharm. Sci. 7(2), 200-4, (2004)

Loading...


Reduced repair capacity of a DNA clustered damage site comprised of 8-oxo-7,8-dihydro-2'-deoxyguanosine and 2-deoxyribonolactone results in an increased mutagenic potential of these lesions. Cunniffe S, O'Neill P, Greenberg MM, et al. Mutat. Res. Fundam. Mol. Mech. Mutagen. 762, 32-9, (2014)

Loading...


Chemoenzymatic synthesis of trans-tetrahydrofuran cores of annonaceous acetogenins from bromobenzene. Ramos JC, Brovetto M, and Seoane GA Org. Lett. 15(8), 1982-5, (2013)

Loading...


Critical chromatography in ternary solvents. Mlynek M and Radke W J. Chromatogr. A 1284, 112-7, (2013)

Loading...


Gymnothelignans A-O: conformation and absolute configuration analyses of lignans bearing tetrahydrofuran from Gymnotheca chinensis. He D, Ding L, Xu H, et al. J. Org. Chem. 77(19), 8435-43, (2012)

Loading...


Metabolism of a serotonin-4 receptor partial agonist 4-{4-[4-tetrahydrofuran-3-yloxy)-benzo[d]isoxazol-3-yloxymethyl]-piperidin-1-ylmethyl}-tetrahydropyran-4-ol (TBPT): identification of an unusual pharmacologically active cyclized oxazolidine metabolite in human. Sawant-Basak A, Coffman KJ, Walker GS, et al. J. Pharm. Sci. 102(9), 3277-93, (2013)

Loading...


Determination of mexiletine in biological fluids by high-performance liquid chromatography. Breithaupt H and Wilfling M J. Chromatogr. A 230(1), 97-105, (1982)

Loading...


Structure of a piperidine-modified calix[4]arene derivative and spectral resolution of its interaction with rare earth metals with chemometric methods. Wang L, Wang X, and Wang Y Spectrochim. Acta. A. Mol. Biomol. Spectrosc. 105, 62-6, (2013)

Loading...


Chemical shifts of phenolic monomers in solution and implications for addition and self-condensation. Haupt RA and Renneckar S Magn. Reson. Chem. 51(2), 95-101, (2013)

Loading...


Determination of heptylphysostigmine in plasma by high-performance liquid chromatography with electrochemical detection. Unni LK and Becker RE J. Chromatogr. A 573(2), 275-81, (1992)

Loading...


Formation of itraconazole-succinic acid cocrystals by gas antisolvent cocrystallization. Ober CA and Gupta RB AAPS PharmSciTech 13(4), 1396-406, (2012)

Loading...


Two new compounds from Schisandra glaucescens. Zhang PP, Gao SS, Zhang TT, et al. J. Asian Nat. Prod. Res. 15(5), 466-72, (2013)

Loading...


Merck 14,9211

Beil. 17,V,1,27

Fieser 1,1140 / Fieser 2,398 / Fieser 3,278 / Fieser 5,649 / Fieser 6,570

Related Products

also commonly purchased with this product

Product #

Description

Add to Cart

Z241296 Whatman® quantitative filter paper, ashless, Grade 40, circles, diam. 125 mm, pack of 100
34860 Methanol, CHROMASOLV®, for HPLC, ≥99.9%
34866 Toluene, CHROMASOLV®, for HPLC, 99.9%
34851 Acetonitrile, CHROMASOLV® gradient grade, for HPLC, ≥99.9%

recommended

Product #

Description

Add to Cart

Z683299 XPell indicating pellets for peroxides prevention, CE for organic solvents

related product

Product #

Description

Add to Cart

Z568066 Pure Solv Micro replacement purification column, activated Alumina
Z568023 Pure Solv Micro 100 Liter solvent purification system, with activated Alumina column, joint Joint: ST/NS 24/40 US

suggested gloves for splash protection

Product #

Description

Add to Cart

Z677639 Butoject butyl gloves, size S
Z677647 Butoject butyl gloves, size M
Z677655 Butoject butyl gloves, size L
Z677663 Butoject butyl gloves, size XL
Z677671 Butoject butyl gloves, size XXL

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?