• USA Home
  • 34920 - Dibutyltin dichloride

EMAIL THIS PAGE TO A FRIEND
34920 Sigma-Aldrich

Dibutyltin dichloride

purum, ≥97.0% (AT)

Synonym: Dibutyldichlorotin

  • CAS Number 683-18-1

  • Linear Formula (CH3CH2CH2CH2)2SnCl2

  • Molecular Weight 303.84

  •  Beilstein Registry Number 3535484

  •  EC Number 211-670-0

  •  MDL number MFCD00000518

  •  PubChem Substance ID 329755089

Purchase

Properties

Related Categories Chemical Synthesis, Organometallic Reagents, Organotin, Organotin Halides
InChI Key   RJGHQTVXGKYATR-UHFFFAOYSA-L
grade   purum
assay   ≥97.0% (AT)
bp   135 °C/10 mmHg(lit.)
mp   37-40 °C(lit.)
  37-41 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Precautionary statements 
RIDADR 
UN 2928 6.1/PG 2
WGK Germany 
3
RTECS 
WH7100000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?