EMAIL THIS PAGE TO A FRIEND
34928 Sigma-Aldrich

Dibutyltin dilaurate

Selectophore

  • CAS Number 77-58-7

  • Linear Formula (CH3CH2CH2CH2)2Sn[OCO(CH2)10CH3]2

  • Molecular Weight 631.56

  •  Beilstein Registry Number 4156980

  •  EC Number 201-039-8

  •  MDL number MFCD00008963

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Plasticizer for ISE More...
grade   Selectophore
vapor pressure   0.2 mmHg ( 160 °C)
InChI Key   UKLDJPRMSDWDSL-UHFFFAOYSA-L
refractive index   n20/D 1.471(lit.)
density   1.066 g/mL at 25 °C(lit.)

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2922 8/PG 3
WGK Germany 
3
RTECS 
WH7000000
Flash Point(F) 
235 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?