• USA Home
  • 35036 - Dichloroacetaldehyde diethyl acetal

EMAIL THIS PAGE TO A FRIEND
35036 Sigma-Aldrich

Dichloroacetaldehyde diethyl acetal

purum, ≥97.0% (GC)

Synonym: 1,1-Dichloro-2,2-diethoxyethane

  • CAS Number 619-33-0

  • Linear Formula Cl2CHCH(OC2H5)2

  • Molecular Weight 187.06

  •  Beilstein Registry Number 1699086

  •  EC Number 210-591-9

  •  MDL number MFCD00000849

Purchase

Properties

InChI Key   CDHLQZJRWKQATP-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.436(lit.)
  n20/D 1.436
bp   183-184 °C(lit.)
density   1.138 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
138.2 °F
Flash Point(C) 
59 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

D54206 Dichloroacetaldehyde diethyl acetal, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?