EMAIL THIS PAGE TO A FRIEND
35190 Sigma-Aldrich

2,5-Dichloroaniline

technical, ≥98.0% (HPLC)

  • CAS Number 95-82-9

  • Linear Formula Cl2C6H3NH2

  • Molecular Weight 162.02

  •  Beilstein Registry Number 1447438

  •  EC Number 202-455-2

  •  MDL number MFCD00007667

  •  PubChem Substance ID 329755119

Purchase

Properties

InChI Key   AVYGCQXNNJPXSS-UHFFFAOYSA-N
grade   technical
assay   ≥98.0% (HPLC)
autoignition temp.   >374 °F
bp   251 °C(lit.)
mp   47-50 °C(lit.)
  48-50 °C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3442 6.1 / PGII
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?