• USA Home
  • 35259 - 2,6-Dichlorobenzaldehyde

EMAIL THIS PAGE TO A FRIEND
35259 Sigma-Aldrich

2,6-Dichlorobenzaldehyde

purum, ≥99.0% (T)

  • CAS Number 83-38-5

  • Linear Formula Cl2C6H3CHO

  • Molecular Weight 175.01

  •  Beilstein Registry Number 386477

  •  EC Number 201-472-2

  •  MDL number MFCD00003307

  •  PubChem Substance ID 329755120

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C7, Carbonyl Compounds, Chemical Synthesis,
InChI Key   DMIYKWPEFRFTPY-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (T)
mp   69-71 °C(lit.)
  69-71 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
2
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

D56500 2,6-Dichlorobenzaldehyde, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?