• USA Home
  • 35390 - 4,4′-Dichlorobenzophenone

EMAIL THIS PAGE TO A FRIEND
35390 Sigma-Aldrich

4,4′-Dichlorobenzophenone

purum, ≥99.0% (GC)

Synonym: 4,4′-DBP

  • CAS Number 90-98-2

  • Linear Formula (ClC6H4)2CO

  • Molecular Weight 251.11

  •  Beilstein Registry Number 643345

  •  EC Number 202-030-1

  •  MDL number MFCD00000623

  •  PubChem Substance ID 24861817

Purchase

Properties

Related Categories Building Blocks, C13 to C14, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   OKISUZLXOYGIFP-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
bp   353 °C(lit.)
mp   144-146 °C(lit.)
  144-147 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DJ0525000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?