• USA Home
  • 356565 - Silver ICP/DCP standard solution

EMAIL THIS PAGE TO A FRIEND
356565 Sigma-Aldrich

Silver ICP/DCP standard solution

10000 μg/mL Ag in 2% HNO3

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, ICP-OES/-MS, ICP/DCP Standard Solutions,
concentration   10000 μg/mL Ag in 2% HNO3
application(s)   ICP/AAS: suitable
density   1.025 g/mL at 25 °C
format   single component solution

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 3264 8/PG 3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?