• USA Home
  • 356719 - Vanadium ICP/DCP standard solution

EMAIL THIS PAGE TO A FRIEND
356719 Sigma-Aldrich

Vanadium ICP/DCP standard solution

10.000 μg/mL V in 1-5% HNO3 (Exact V concentration on label)

Purchase

Properties

Related Categories ICP/DCP Standard Solutions, Materials Science, Reference/Calibration Standards, Standard Solutions
contains   hydrazine as stabilizer (trace)
concentration   10.000 μg/mL V in 1-5% HNO3 (Exact V concentration on label)
application(s)   ICP/AAS: suitable
density   1.035 g/mL at 25 °C
format   single component solution

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2031B 8 / PGII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?