• USA Home
  • 356743 - Zinc ICP/DCP standard solution

EMAIL THIS PAGE TO A FRIEND
356743 Sigma-Aldrich

Zinc ICP/DCP standard solution

analytical standard, 10,000 μg/mL Zn+2 in 2% HNO3 (Exact Zn concentration on label)

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Elemental & Metal Standards, Elemental, Ions and Water (Karl Fischer) Standards, ICP/DCP Standard Solutions,
grade   analytical standard
concentration   10,000 μg/mL Zn+2 in 2% HNO3 (Exact Zn concentration on label)
application(s)   ICP/AAS: suitable
density   1.03 g/mL at 25 °C
format   single component solution

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3264 8 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?