• USA Home
  • 356751 - Zirconium ICP/DCP standard solution

EMAIL THIS PAGE TO A FRIEND
356751 Sigma-Aldrich

Zirconium ICP/DCP standard solution

analytical standard, 10,000 μg/mL Zr in 4 wt. % HCl (Exact Zr concentration on label)

Synonym: Zirconium solution

  • Empirical Formula (Hill Notation) Zr

  • Molecular Weight 91.22

  •  MDL number MFCD00011303

  •  PubChem Substance ID 329755201

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Elemental & Metal Standards, Elemental, Ions and Water (Karl Fischer) Standards, Food and Cosmetic Component Standards,
grade   analytical standard
InChI Key   QCWXUUIWCKQGHC-UHFFFAOYSA-N
concentration   10,000 μg/mL Zr in 4 wt. % HCl (Exact Zr concentration on label)
application(s)   ICP/AAS: suitable
density   1.01 g/mL at 25 °C
format   single component solution

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1789 8 / PGIII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?