EMAIL THIS PAGE TO A FRIEND
35824 Sigma-Aldrich

3-Chloroaniline

PESTANAL®, analytical standard

  • CAS Number 108-42-9

  • Linear Formula ClC6H4NH2

  • Molecular Weight 127.57

  •  Beilstein Registry Number 605969

  •  EC Number 203-581-0

  •  MDL number MFCD00007765

  •  PubChem Substance ID 24861962

Purchase

Properties

Related Categories 2005/70/EU, Additional Standards, Alpha Index of All Pesticide Standards, Amines, Analytical Standards,
grade   analytical standard
vapor pressure   1 mmHg ( 63.5 °C)
InChI Key   PNPCRKVUWYDDST-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.594(lit.)
bp   95-96 °C/11 mmHg(lit.)
mp   −11-−9 °C(lit.)
density   1.206 g/mL at 25 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2019 6.1 / PGII
WGK Germany 
3
RTECS 
BX0350000
Flash Point(F) 
244.4 °F
Flash Point(C) 
118 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?