• USA Home
  • 35825 - Dichloroacetic anhydride

EMAIL THIS PAGE TO A FRIEND
35825 Sigma-Aldrich

Dichloroacetic anhydride

technical, ≥94% (T)

  • CAS Number 4124-30-5

  • Linear Formula (Cl2CHCO)2O

  • Molecular Weight 239.87

  •  Beilstein Registry Number 512173

  •  EC Number 223-924-8

  •  MDL number MFCD00000842

Purchase

Properties

InChI Key   RQHMQURGSQBBJY-UHFFFAOYSA-N
grade   technical
assay   ≥94% (T)
refractive index   n20/D 1.483(lit.)
  n20/D 1.483
bp   214-216 °C(lit.)
mp   ~30 °C
density   1.574 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2923 6.1(8) / PGIII
WGK Germany 
3
RTECS 
AG6300000
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

331295 Dichloroacetic anhydride, technical grade, 85%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?