• USA Home
  • 35853 - 1,5-Dichloro-1,1,3,3,5,5-hexamethyltrisiloxane

EMAIL THIS PAGE TO A FRIEND
35853 Sigma-Aldrich

1,5-Dichloro-1,1,3,3,5,5-hexamethyltrisiloxane

technical, ≥95% (GC)

Synonym: 1,5-Dichloro-hexamethyltrisiloxane

  • CAS Number 3582-71-6

  • Linear Formula [ClSi(CH3)2O]2Si(CH3)2

  • Molecular Weight 277.37

  •  Beilstein Registry Number 1763185

  •  EC Number 222-707-5

  •  MDL number MFCD00045147

  •  PubChem Substance ID 329755230

Purchase

Properties

InChI Key   GJIYNWRLGOMDEX-UHFFFAOYSA-N
grade   technical
assay   ≥95% (GC)
refractive index   n20/D 1.405(lit.)
  n20/D 1.406
bp   184 °C(lit.)
density   1.018 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2987 8 / PGII
WGK Germany 
3
Flash Point(F) 
168.8 °F
Flash Point(C) 
76 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?