• USA Home
  • 35975 - 1,2-Dibromo-1-chloroethane

EMAIL THIS PAGE TO A FRIEND
35975 Sigma-Aldrich

1,2-Dibromo-1-chloroethane

analytical standard, for the determination of vinyl chloride

  • CAS Number 598-20-9

  • Linear Formula CHBrClCH2Br

  • Molecular Weight 222.31

  •  Beilstein Registry Number 1731853

  •  MDL number MFCD00142465

  •  PubChem Substance ID 329755239

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Bromo,
grade   analytical standard
InChI Key   DVTKTUMRXCKEEG-UHFFFAOYSA-N
form   neat
quality   for the determination of vinyl chloride
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?