• USA Home
  • 35996 - 2,4,6-Trichloroaniline

EMAIL THIS PAGE TO A FRIEND
35996 Sigma-Aldrich

2,4,6-Trichloroaniline

PESTANAL®, analytical standard

Purchase

Properties

Related Categories Additional Standards, Alpha Index of All Pesticide Standards, Amines, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   NATVSFWWYVJTAZ-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   262 °C(lit.)
mp   73-75 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN2811 - class 6.1 - PG 3 - EHS - Toxic solids, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
BZ0250000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?