• USA Home
  • 35998 - 2,4,6-Trimethylphenol

EMAIL THIS PAGE TO A FRIEND
35998 Sigma-Aldrich

2,4,6-Trimethylphenol

PESTANAL®, analytical standard

Synonym: Mesitol

  • CAS Number 527-60-6

  • Linear Formula (CH3)3C6H2OH

  • Molecular Weight 136.19

  •  Beilstein Registry Number 1859675

  •  EC Number 208-419-2

  •  MDL number MFCD00002235

  •  PubChem Substance ID 329755248

Purchase

Properties

Related Categories Additional Standards, All Alphabetically Sorted, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   BPRYUXCVCCNUFE-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   220 °C(lit.)
mp   70-72 °C(lit.)
  71-74 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN2430 - class 8 - PG 3 - EHS - Alkylphenols, solid, n.o.s., HI: A,S
WGK Germany 
2
RTECS 
OX6590000
Flash Point(F) 
208.4 °F
Flash Point(C) 
98 °C
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?