• USA Home
  • 36000 - 5,6-Dichloronicotinic acid

EMAIL THIS PAGE TO A FRIEND
36000 Sigma-Aldrich

5,6-Dichloronicotinic acid

purum, ≥98.0% (HPLC)

Synonym: 5,6-Dichloropyridine-3-carboxylic acid

  • CAS Number 41667-95-2

  • Empirical Formula (Hill Notation) C6H3Cl2NO2

  • Molecular Weight 192.00

  •  Beilstein Registry Number 383740

  •  MDL number MFCD00075181

  •  PubChem Substance ID 329755250

Purchase

Properties

Related Categories Building Blocks, C6, Chemical Synthesis, Halogenated Heterocycles, Heterocyclic Building Blocks,
InChI Key   RNRLTTNKVLFZJS-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
mp   163-167 °C
  164-168 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?