360465 Sigma-Aldrich

1-Butanol

ACS reagent, ≥99.4%

DOWNLOAD MSDS (PDF)

Synonym: n-Butanol, Butyl alcohol

Properties

Related Categories ACS Grade, ACS Grade Solvents, ACS and Reagent Grade Solvents, Amber Glass Bottles, Carbon Steel Flex-Spout Cans,
grade   ACS reagent
vapor density   2.55 (vs air)
assay   ≥99.4%
autoignition temp.   649 °F
expl. lim.   11.2 %
impurities   ≤0.0008 meq/g Titr. acid
  ≤0.01% butyraldehyde
  ≤0.1% water
  ≤0.2% butyl ether
evapn. residue   ≤0.005%
color   APHA: ≤10
refractive index   n20/D 1.399(lit.)
bp   116-118 °C(lit.)
mp   −90 °C(lit.)
density   0.81 g/mL at 25 °C(lit.)

Description

Packaging

1, 2, 2.5, 4×2.5, 4×4 L in glass bottle

20 L in steel drum

500 mL in glass bottle

Price and Availability

Customers Also Viewed

1-Butanol

for molecular biology, ≥99%

1-Butanol

analytical standard

1-Butanol
2815491-Butanol

anhydrous, 99.8%

1-Butanol

CHROMASOLV® Plus, for HPLC, ≥99.7%

1-Propanol

ACS reagent, ≥99.5%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?