360503 Sigma-Aldrich

2-Methoxyethanol

contains 50 ppm BHT as stabilizer, ACS reagent, ≥99.3%

DOWNLOAD MSDS (PDF)

Synonym: Ethylene glycol monomethyl ether, Methyl glycol

Properties

Related Categories ACS Grade, ACS Grade Solvents, ACS and Reagent Grade Solvents, Amber Glass Bottles, Carbon Steel Flex-Spout Cans,
grade   ACS reagent
vapor density   2.62 (vs air)
vapor pressure   6.17 mmHg ( 20 °C)
assay   ≥99.3%
autoignition temp.   548 °F
contains   50 ppm BHT as stabilizer
expl. lim.   24.5 %
impurities   ≤0.002 meq/g Titr. acid
  ≤0.100% water
color   APHA: ≤10
refractive index   n20/D 1.402(lit.)
bp   124-125 °C(lit.)
mp   −85 °C(lit.)
density   0.965 g/mL at 25 °C(lit.)

Description

Packaging

1, 2.5, 4×4 L in glass bottle

20 L in steel drum

500 mL in glass bottle

Price and Availability

Customers Also Viewed

2-Methoxyethanol

anhydrous, 99.8%

2-Methoxyethanol

CHROMASOLV®, for HPLC, ≥99.9%

2-Methoxyethanol

ReagentPlus®, ≥99.0%, contains 50 ppm BHT as stabilizer

2-Methoxyethanol

for amino acid analysis, ≥99.5% (GC)

2-Methoxyethanol

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?