CAS Number 110-86-1
Empirical Formula (Hill Notation) C5H5N
Molecular Weight 79.10
Beilstein Registry Number 103233
EC Number 203-809-9
MDL number MFCD00011732
PubChem Substance ID 24862172
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | ACS Grade, ACS Grade Solvents, ACS and Reagent Grade Solvents, Amber Glass Bottles, Carbon Steel Cans with NPT Threads, |
| grade | ACS reagent |
| vapor density | 2.72 (vs air) |
| vapor pressure | 10 mmHg ( 13.2 °C) |
| 20 mmHg ( 25 °C) | |
| assay | ≥99.0% |
| autoignition temp. | 899 °F |
| expl. lim. | 12.4 % |
1, 2, 2.5, 4×4 L in glass bottle
20 L in steel drum
View returnable container options.
100, 500, 6×500 mL in glass bottle
200, 400 L in Pure-Pac™ 1
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?