EMAIL THIS PAGE TO A FRIEND
36140 Sigma-Aldrich

2,5-Dichlorophenol

purum, ≥98.0% (HPLC)

  • CAS Number 583-78-8

  • Linear Formula Cl2C6H3OH

  • Molecular Weight 163.00

  •  Beilstein Registry Number 1907692

  •  EC Number 209-520-4

  •  MDL number MFCD00002174

  •  PubChem Substance ID 24862219

Purchase

Properties

Related Categories Building Blocks, C6 to C8, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds,
InChI Key   RANCECPPZPIPNO-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
form   crystals
  powder
bp   211 °C(lit.)
mp   54-57 °C(lit.)
  54-57 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2020 6.1 / PGIII
WGK Germany 
2
RTECS 
SK8600000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?