• USA Home
  • 36214 - 4,5-Dichloro-o-phenylenediamine

EMAIL THIS PAGE TO A FRIEND
36214 Sigma-Aldrich

4,5-Dichloro-o-phenylenediamine

technical, ≥95.0% (AT)

Synonym: 1,2-Diamino-4,5-dichlorobenzene, 4,5-Dichloro-1,2-phenylenediamine

  • CAS Number 5348-42-5

  • Linear Formula Cl2C6H2(NH2)2

  • Molecular Weight 177.03

  •  Beilstein Registry Number 1072811

  •  EC Number 226-305-0

  •  MDL number MFCD00007723

  •  PubChem Substance ID 24862292

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks, Polyamines More...
InChI Key   IWFHBRFJOHTIPU-UHFFFAOYSA-N
grade   technical
assay   ≥95.0% (AT)
  ≥97.0% (GC)
mp   158-164 °C(lit.)
  158-164 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?