• USA Home
  • 36242 - Phenylphosphonic dichloride

EMAIL THIS PAGE TO A FRIEND
36242 Sigma-Aldrich

Phenylphosphonic dichloride

technical, ≥90% (GC)

Synonym: P,P-Dichlorophenylphosphine oxide

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Phosphine Compounds, Phosphorus Precursors
InChI Key   IBDMRHDXAQZJAP-UHFFFAOYSA-N
grade   technical
assay   ≥90% (GC)
refractive index   n20/D 1.559(lit.)
  n20/D 1.559
bp   258 °C(lit.)
mp   3 °C(lit.)
density   1.375 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 3265 8 / PGII
WGK Germany 
3
Flash Point(F) 
399.2 °F
Flash Point(C) 
204 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

389560 Phenylphosphonic dichloride, technical grade, 90%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?