EMAIL THIS PAGE TO A FRIEND
36260 Sigma-Aldrich

1,3-Dichloropropane

purum, ≥98.0% (GC)

Synonym: Trimethylene dichloride

Purchase

Properties

Related Categories Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
InChI Key   YHRUOJUYPBUZOS-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.448(lit.)
  n20/D 1.448
bp   120-122 °C(lit.)
mp   −99 °C(lit.)
density   1.19 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
2
RTECS 
TX9660000
Flash Point(F) 
96.8 °F
Flash Point(C) 
36 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

D72204 1,3-Dichloropropane, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?