• USA Home
  • 36290 - 1,3-Dichloro-2-propanol

EMAIL THIS PAGE TO A FRIEND
36290 Sigma-Aldrich

1,3-Dichloro-2-propanol

purum, ≥98.0% (GC)

Synonym: Glycerol-α,γ-dichlorohydrin

  • CAS Number 96-23-1

  • Linear Formula (ClCH2)2CHOH

  • Molecular Weight 128.99

  •  Beilstein Registry Number 1732063

  •  EC Number 202-491-9

  •  MDL number MFCD00000951

Purchase

Properties

Related Categories Alcohols, Building Blocks, C2 to C6, Chemical Synthesis, Organic Building Blocks,
InChI Key   DEWLEGDTCGBNGU-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.483(lit.)
  n20/D 1.484
bp   174.3 °C(lit.)
mp   −4 °C(lit.)
density   1.363 g/mL at 20 °C
  1.351 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2750 6.1 / PGII
WGK Germany 
3
RTECS 
UB1400000
Flash Point(F) 
186.8 °F
Flash Point(C) 
86 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?