• USA Home
  • 36355 - 2,3-Dichloropropionyl chloride

EMAIL THIS PAGE TO A FRIEND
36355 Sigma-Aldrich

2,3-Dichloropropionyl chloride

purum, ≥97.0% (GC)

  • CAS Number 7623-13-4

  • Linear Formula CH2ClCHClCOCl

  • Molecular Weight 161.41

  •  Beilstein Registry Number 506750

  •  EC Number 231-541-2

  •  MDL number MFCD00000713

Purchase

Properties

Related Categories Acid Halides, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks More...
InChI Key   JQELECXPPAOSTM-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.477(lit.)
  n20/D 1.477
bp   53-54 °C/17 mmHg(lit.)
density   1.476 g/mL at 25 °C(lit.)

Description

Other Notes

Precursor of 2-chloroacryloyl chloride a ketene equivalent. Review

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 3265 8 / PGII
WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?