EMAIL THIS PAGE TO A FRIEND
36390 Sigma-Aldrich

2,6-Dichloropurine

purum, ≥97.0% (AT)

  • CAS Number 5451-40-1

  • Empirical Formula (Hill Notation) C5H2Cl2N4

  • Molecular Weight 189.00

  •  Beilstein Registry Number 9354

  •  EC Number 226-681-6

  •  MDL number MFCD00077725

  •  PubChem Substance ID 329755345

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Halogenated Heterocycles, Heterocyclic Building Blocks, Purines More...
InChI Key   RMFWVOLULURGJI-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (AT)
mp   185-195 °C (dec.)(lit.)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?