EMAIL THIS PAGE TO A FRIEND
36398 Sigma-Aldrich

2,3-Dichloropyridine

purum, ≥97.0% (AT)

  • CAS Number 2402-77-9

  • Empirical Formula (Hill Notation) C5H3Cl2N

  • Molecular Weight 147.99

  •  Beilstein Registry Number 109811

  •  EC Number 219-281-8

  •  MDL number MFCD00006229

  •  PubChem Substance ID 329755347

Purchase

Properties

InChI Key   MAKFMOSBBNKPMS-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (AT)
mp   64-67 °C(lit.)
  64-67 °C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
US8050000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

D73405 2,3-Dichloropyridine, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?