• USA Home
  • 36445 - Heating oil test marker 2

EMAIL THIS PAGE TO A FRIEND
36445 Sigma-Aldrich

Heating oil test marker 2

rapid analysis of colored markers in heating oil

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Application Specific Reagents, Heating Oil Test
grade   rapid analysis of colored markers in heating oil

Description

Packaging

Carton with 20 test tubes

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1789 8 / PGII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?