EMAIL THIS PAGE TO A FRIEND
36502 Sigma-Aldrich

Dimefox

PESTANAL®, analytical standard

Synonym: Tetramethylphosphorodiamidic fluoride

  • CAS Number 115-26-4

  • Empirical Formula (Hill Notation) C4H12FN2OP

  • Molecular Weight 154.12

  •  Beilstein Registry Number 1704257

  •  EC Number 204-076-8

  •  MDL number MFCD00055417

Purchase

Properties

Related Categories Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, D, Insecticides,
grade   analytical standard
InChI Key   PGJBQBDNXAZHBP-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1 / PGI
WGK Germany 
2
RTECS 
TD4025000
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?