EMAIL THIS PAGE TO A FRIEND
36516 Sigma-Aldrich

Atraton-desisopropyl

PESTANAL®, analytical standard

Synonym: 2-Amino-4-ethylamino-6-methoxy-1,3,5-triazine

  • CAS Number 30360-56-6

  • Empirical Formula (Hill Notation) C6H11N5O

  • Molecular Weight 169.18

  •  Beilstein Registry Number 8453

  •  MDL number MFCD00142858

  •  PubChem Substance ID 329755368

Purchase

Properties

Related Categories A-B, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, Herbicides,
grade   PESTANAL®, analytical standard
InChI Key   MZOHHVOQVUFCIY-UHFFFAOYSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?