• USA Home
  • 36571 - 3,4-Dichlorobenzoic acid

EMAIL THIS PAGE TO A FRIEND
36571 Sigma-Aldrich

3,4-Dichlorobenzoic acid

PESTANAL®, analytical standard

  • CAS Number 51-44-5

  • Linear Formula Cl2C6H3CO2H

  • Molecular Weight 191.01

  •  Beilstein Registry Number 2044777

  •  EC Number 200-099-2

  •  MDL number MFCD00002492

  •  PubChem Substance ID 329755402

Purchase

Properties

Related Categories Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, D, Pesticides & Metabolites Standards More...
grade   PESTANAL®, analytical standard
InChI Key   VPHHJAOJUJHJKD-UHFFFAOYSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   204-206 °C(lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DG7175000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?