• USA Home
  • 36586 - α,α′-Dichloro-p-xylene

EMAIL THIS PAGE TO A FRIEND
36586 Sigma-Aldrich

α,α′-Dichloro-p-xylene

purum, ≥98.0% (GC)

Synonym: 1,4-Bis(chloromethyl)benzene, p-Xylylene dichloride

Purchase

Properties

Related Categories Aryl, Building Blocks, C8, Chemical Synthesis, Halogenated Hydrocarbons,
InChI Key   ZZHIDJWUJRKHGX-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
bp   254 °C(lit.)
mp   98-101 °C(lit.)
  98-101 °C
solubility   methanol: 50 mg/mL clear
storage temp.   2-8°C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN3077 - class 9 - PG 3 - DOT/IATA UN3335 - Environmentally hazardous substances, solid, n.o.s., HI: all (not BR)
WGK Germany 
3
RTECS 
ZE4100000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

105740 α,α′-Dichloro-p-xylene, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?