• USA Home
  • 36589 - Terbuthylazin solution

EMAIL THIS PAGE TO A FRIEND
36589 Sigma-Aldrich

Terbuthylazin solution

100 ng/μL in methanol, PESTANAL®, analytical standard

  • CAS Number 5915-41-3

  • Empirical Formula (Hill Notation) C9H16ClN5

  • Molecular Weight 229.71

  •  Beilstein Registry Number 3951137

  •  EC Number 200-659-6

  •  MDL number MFCD00055513

  •  PubChem Substance ID 329755412

Purchase

Properties

Related Categories Additional Standards, All Herbicide Standards, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   FZXISNSWEXTPMF-UHFFFAOYSA-N
product line   PESTANAL®
shelf life   limited shelf life, expiry date on the label
concentration   100 ng/μL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

General description

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706023

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1230 3/PG 2
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide, Modified Gradient

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide, Modified Gradient
Keywords: Chromatography, High performance liquid chromatography, Pesticides, Separation

HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide
Keywords: Chromatography, High performance liquid chromatography, Pesticides, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?