• USA Home
  • 36614 - Dimethyl 5-nitroisophthalate

EMAIL THIS PAGE TO A FRIEND
36614 Sigma-Aldrich

Dimethyl 5-nitroisophthalate

PESTANAL®, analytical standard

Synonym: Dimethyl 5-nitrobenzene-1,3-dicarboxylate

  • CAS Number 13290-96-5

  • Linear Formula O2NC6H3-1,3-(CO2CH3)2

  • Molecular Weight 239.18

  •  Beilstein Registry Number 2140916

  •  EC Number 236-307-3

  •  MDL number MFCD00008429

  •  PubChem Substance ID 24862618

Purchase

Properties

Related Categories Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, D, Pesticides & Metabolites Standards More...
grade   PESTANAL®, analytical standard
InChI Key   GGTSJKFPGKFLCZ-UHFFFAOYSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   123-125 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
CZ4340000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?