EMAIL THIS PAGE TO A FRIEND
36662 Sigma-Aldrich

4,4′-DDT solution

100 ng/μL in methanol, PESTANAL®, analytical standard

Synonym: 1,1,1-Trichloro-2,2-bis(4-chlorophenyl)ethane solution, 4,4′-DDT solution

  • CAS Number 50-29-3

  • Empirical Formula (Hill Notation) C14H9Cl5

  • Molecular Weight 354.49

  •  Beilstein Registry Number 1882657

  •  EC Number 200-659-6

  •  MDL number MFCD00000802

  •  PubChem Substance ID 329755436

Purchase

Properties

Related Categories 2000/60/EC, Additional Standards, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   YVGGHNCTFXOJCH-UHFFFAOYSA-N
product line   PESTANAL®
shelf life   limited shelf life, expiry date on the label
concentration   100 ng/μL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C
Gene Information   mouse ... Esr1(13982)
rat ... Ar(24208)

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

31041 4,4′-DDT, PESTANAL®, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?