EMAIL THIS PAGE TO A FRIEND
36683 Sigma-Aldrich

2-Aminophenol

PESTANAL®, analytical standard

  • CAS Number 95-55-6

  • Linear Formula H2NC6H4OH

  • Molecular Weight 109.13

  •  Beilstein Registry Number 606075

  •  EC Number 202-431-1

  •  MDL number MFCD00007690

  •  PubChem Substance ID 329755449

Purchase

Properties

Related Categories A-B, Additional Standards, Alpha Index of All Pesticide Standards, Amines, Analytical Standards,
grade   analytical standard
InChI Key   CDAWCLOXVUBKRW-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   170-175 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2512 6.1 / PGIII
WGK Germany 
3
RTECS 
SJ4950000
Flash Point(F) 
334.4 °F
Flash Point(C) 
168 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?