EMAIL THIS PAGE TO A FRIEND
36689 Sigma-Aldrich

Azobenzene

PESTANAL®, analytical standard

  • CAS Number 103-33-3

  • Linear Formula C6H5N=NC6H5

  • Molecular Weight 182.22

  •  Beilstein Registry Number 1819138

  •  EC Number 203-102-5

  •  MDL number MFCD00003022

  •  PubChem Substance ID 24862720

Purchase

Properties

Related Categories A-B, Acaricides, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
vapor pressure   1 mmHg ( 104 °C)
InChI Key   DMLAVOWQYNRWNQ-BUHFOSPRSA-N
product line   PESTANAL®
form   neat
autoignition temp.   890 °F
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   293 °C(lit.)
mp   65-68 °C(lit.)
density   1.09 g/mL at 25 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
CN1400000
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?