EMAIL THIS PAGE TO A FRIEND
36690 Sigma-Aldrich

4,4′-Bipyridyl

anhydrous, PESTANAL®, analytical standard

Synonym: 4,4′-Bipyridine

  • CAS Number 553-26-4

  • Empirical Formula (Hill Notation) C10H8N2

  • Molecular Weight 156.18

  •  Beilstein Registry Number 113176

  •  EC Number 209-036-3

  •  MDL number MFCD00006416

  •  PubChem Substance ID 329755454

Purchase

Properties

Related Categories A-B, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, Pesticides & Metabolites Standards More...
grade   analytical standard
InChI Key   MWVTWFVJZLCBMC-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
quality   anhydrous
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   305 °C(lit.)
mp   109-112 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
DW1760000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?